Learn More
Chemglass Life Sciences 0-12mm, 90°, Glass Only
SDP
Catalog No. 501532014
Encompass_Preferred
0-12mm, 90, Glass Only. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.
Catalog No. 50-153-2014 Supplier Chemglass Life Sciences Supplier No. CGK50303
0-12mm, 90, Glass Only. Hi-vac valve with two sidearms sealed to the barrel at a 90 angle to one another. Designed with a precision external thread for fine control.