Learn More
Chemglass Life Sciences 0-12mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum, Perfluoro O-Ring
SDP
Catalog No. 501531871
Encompass_Preferred
0-12mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum, Perfluoro O-Ring. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.
Catalog No. 50-153-1871 Supplier Chemglass Life Sciences Supplier No. CG96153
0-12mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum, Perfluoro O-Ring. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.