Learn More
Chemglass Life Sciences 0-14mm Stem and Control Knob, Blue Band Indicator for Low Temperature Applications
SDP
Catalog No. 50872696
Encompass_Preferred
0-14mm Stem and Control Knob, Blue Band Indicator for Low Temperature Applications. Replacement one-piece stem and control knob for our NEW 0-14mm drain valve. Available with either a red indicator band used for high temperature applications up to 230°C, or with a blue indicator band used for low temperature applications down to -60°C. The high temperature plug uses Perfluoro o-rings on shaft and tip, while the low temperature plug uses Perfluoro o-rings on shaft and a FEP-Silicone o-ring on the tip.
Catalog No. 50-872-696 Supplier Chemglass Life Sciences Supplier No. CG911A11
0-14mm Stem and Control Knob, Blue Band Indicator for Low Temperature Applications. Replacement one-piece stem and control knob for our NEW 0-14mm drain valve. Available with either a red indicator band used for high temperature applications up to 230°C, or with a blue indicator band used for low temperature applications down to -60°C. The high temperature plug uses Perfluoro o-rings on shaft and tip, while the low temperature plug uses Perfluoro o-rings on shaft and a FEP-Silicone o-ring on the tip.