Learn More
Chemglass Life Sciences 0-14mm Stem and Control Knob, White Band Indicator, Spring Loaded
SDP
Supplier:  Chemglass Life Sciences CG911X10
Encompass_Preferred
Replacement one-piece stem and control knob for our NEW 0-14mm drain valve. Available with either a red indicator band used for high temperature applications up to 230C, or with a blue indicator band used for low temperature applications down to -60C, or with a white band indicator which is spring loaded and uses the same o-ring as the high temperure valve. The high temperature plug uses Perfluoro o-rings on shaft and tip, while the low temperature plug uses Perfluoro o-rings on shaft and a FEP-Silicone o-ring on the tip.
Catalog No. 50-009-80722