Learn More
Fine Science Tools Minutien Pins, Tip: 0.0175 mm, 1 cm, 500/pk
SDP
Catalog No. NC9679721
Encompass
Minutien Pins - Black Anodized/0.15mm Diameter/ Tip Diameter: 0.0175mm/ Rod Diameter: 0.15mm/ Alloy: Black Anodized Steel/ Length: 1cm/ Unit: 500
Catalog No. NC9679721 Supplier Fine Science Tools Supplier No. 2600215
Minutien Pins - Black Anodized/0.15mm Diameter/ Tip Diameter: 0.0175mm/ Rod Diameter: 0.15mm/ Alloy: Black Anodized Steel/ Length: 1cm/ Unit: 500