Learn More
Ace Glass, Inc. 0.16" 316 stainless steel packing
SDP
Catalog No. 509557366
Encompass
0.16 316 stainless steel PRO-PAK distillation packing. An efficient protruded metal dump-packing for distillation columns of 0.75 to 12 diameter. Suitable for both atmospheric and reduced pressures down to 10mm Hg condenser pressure.
Catalog No. 50-955-7366 Supplier Ace Glass, Inc. Supplier No. 662404
0.16 316 stainless steel PRO-PAK distillation packing. An efficient protruded metal dump-packing for distillation columns of 0.75 to 12 diameter. Suitable for both atmospheric and reduced pressures down to 10mm Hg condenser pressure.