Learn More
Inorganic Ventures 0.18 M Sodium Carbonate 0.17
SDP
Catalog No. 50723542
Encompass_Preferred
Inorganic Ventures' 0.18 M Sodium Carbonate/ 0.17 M Sodium Bicarbonate is a 500mL reference material set in a water matrix for stability. This fluoride eluent is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-542 Supplier Inorganic Ventures Supplier No. ELUENT1817500ML
Inorganic Ventures' 0.18 M Sodium Carbonate/ 0.17 M Sodium Bicarbonate is a 500mL reference material set in a water matrix for stability. This fluoride eluent is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search