Learn More
Teknova 0.1M Citric Acid Solution. 700mL, Sterile. 4 pack.
SDP
Catalog No. 502387313
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7313 Supplier Teknova Supplier No. C24404PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.