Learn More
Hach Company Conductivity Standard Solution, 12890 µS/cm, KCl, 500 mL
Catalog No. 501996985 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. 12890 µS/cm ±10% at 25°C, 7455 mg/L ±10% as Potassium Chloride (KCl) (0.1M). 500 mL bottle.
Catalog No. 50-199-6985 Supplier Hach Company Supplier No. 2974449
Conductivity Standard Solution. 12890 µS/cm ±10% at 25°C, 7455 mg/L ±10% as Potassium Chloride (KCl) (0.1M). 500 mL bottle.