Learn More
Teknova 0.1M Sodium Citrate, pH 2.8 , 4L, Endotoxin Tested. Sterile. 4 pack.
SDP
Catalog No. 502745817
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-274-5817 Supplier Teknova Supplier No. S09284PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.