Learn More
Teknova 0.1M Sodium Citrate, pH 5.0. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387380
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7380 Supplier Teknova Supplier No. S02312PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.