Learn More
Genesee Scientific Corporation Genesee Scientific 0.1ml 8-Well PCR Strip Tubes, Ultra-Thin Walls, Individually Attached Caps, 120 Strips/Unit
SDP
Catalog No. NC2400214
Encompass
The Genesee Scientific 27-125UL are 0.1ml 8-well PCR strip tubes that enhance molecular biology applications, particularly real-time PCR (qPCR). Designed with individually attached, hinged clear flat caps opening at a 45º angle, they ensure easy access without compromising the PCR rack space. Made from high-quality virgin polypropylene with ultra-thin walls for superior heat transfer, these tubes guarantee reliable and reproducible results, certified free from contaminants like RNase, DNase, and pyrogens.
Catalog No. NC2400214 Supplier Genesee Scientific Corporation Supplier No. 27125UL
The Genesee Scientific 27-125UL are 0.1ml 8-well PCR strip tubes that enhance molecular biology applications, particularly real-time PCR (qPCR). Designed with individually attached, hinged clear flat caps opening at a 45º angle, they ensure easy access without compromising the PCR rack space. Made from high-quality virgin polypropylene with ultra-thin walls for superior heat transfer, these tubes guarantee reliable and reproducible results, certified free from contaminants like RNase, DNase, and pyrogens.