Learn More
Chemglass Life Sciences 0.1 mL 96-Well PCR Plate, No Skirt, White, H12 Notch (Nest, CGN-3617-D-01)
SDP
Catalog No. 502733418
Encompass_Preferred
This 0.1 mL, 96-well PCR plate is designed for low-volume PCR applications. It features a no-skirt format and an H12 orientation notch, and is supplied in packs of 25. Plates are RNase/DNase/pyrogen free and autoclavable, suitable for high-throughput thermal cycling.
  • Material: polypropylene.
  • Well volume: 0.1 mL.
  • Well count: 96 wells.
  • Skirt type: no skirt.
  • Color: white.
  • Notch position: H12.
  • Pack quantity: packs of 25.
  • RNase/DNase/pyrogen free.
  • Autoclavable.
Catalog No. 50-273-3418 Supplier Chemglass Life Sciences Supplier No. CGN3617D01
This 0.1 mL, 96-well PCR plate is designed for low-volume PCR applications. It features a no-skirt format and an H12 orientation notch, and is supplied in packs of 25. Plates are RNase/DNase/pyrogen free and autoclavable, suitable for high-throughput thermal cycling.
  • Material: polypropylene.
  • Well volume: 0.1 mL.
  • Well count: 96 wells.
  • Skirt type: no skirt.
  • Color: white.
  • Notch position: H12.
  • Pack quantity: packs of 25.
  • RNase/DNase/pyrogen free.
  • Autoclavable.