Learn More
Chemglass Life Sciences Chemglass 0.1 mL 96-Well PCR Plate, Semi-Skirt, Clear, A1 Notch, 25/Pack
SDP
Catalog No. 502733419
Encompass_Preferred
The Chemglass 0.1 mL 96-well PCR plate (Nest line) is a clear, semi-skirted polypropylene plate engineered for PCR and qPCR workflows. It offers 0.1 mL well volume, RNase/DNase/pyrogen-free surfaces, and autoclavability for reliable molecular biology performance. The plate's A1 notch and semi-skirt design support correct orientation and compatibility with automated handlers and standard thermal cyclers.
  • Provides 0.1 mL well volume for low-reaction-volume assays.
  • Features 96 wells in the standard microplate format.
  • Semi-skirted design balances rigidity with a compact footprint for automation.
  • Includes an A1 notch for positive plate orientation.
  • Made from clear polypropylene for optical clarity and chemical resistance.
  • Manufactured RNase/DNase/pyrogen free to protect nucleic acid integrity.
  • Autoclavable for decontamination and reuse.
  • Packaged 25 plates per pack to support batch processing workflows.
Catalog No. 50-273-3419 Supplier Chemglass Life Sciences Supplier No. CGN3617C01
Add to Cart
Add to Cart
The Chemglass 0.1 mL 96-well PCR plate (Nest line) is a clear, semi-skirted polypropylene plate engineered for PCR and qPCR workflows. It offers 0.1 mL well volume, RNase/DNase/pyrogen-free surfaces, and autoclavability for reliable molecular biology performance. The plate's A1 notch and semi-skirt design support correct orientation and compatibility with automated handlers and standard thermal cyclers.
  • Provides 0.1 mL well volume for low-reaction-volume assays.
  • Features 96 wells in the standard microplate format.
  • Semi-skirted design balances rigidity with a compact footprint for automation.
  • Includes an A1 notch for positive plate orientation.
  • Made from clear polypropylene for optical clarity and chemical resistance.
  • Manufactured RNase/DNase/pyrogen free to protect nucleic acid integrity.
  • Autoclavable for decontamination and reuse.
  • Packaged 25 plates per pack to support batch processing workflows.