Learn More
Chemglass Life Sciences Chemglass 0.1 mL 96-Well Semi-Skirt PCR Plate, White, A1 Notch
SDP
Catalog No. 502733415
Encompass_Preferred
Low-volume 0.1 mL, 96-well PCR plate engineered for efficient heat transfer and reliable thermal cycling. Thin-walled polypropylene construction delivers chemical compatibility and low sample loss, and plates are DNase-, RNase-, and pyrogen-free for sensitive molecular biology applications.
  • Well volume: 0.1 mL.
  • Wells: 96.
  • Skirt type: Semi skirt.
  • Color: White.
  • Notch position: A1 notch.
  • Pack quantity: 25 per pack.
  • Material: Polypropylene.
  • DNase- and RNase-free, and pyrogen free.
  • Autoclavable.
  • Typical application: PCR and thermal cycling workflows.
Catalog No. 50-273-3415 Supplier Chemglass Life Sciences Supplier No. CGN3617W01
Low-volume 0.1 mL, 96-well PCR plate engineered for efficient heat transfer and reliable thermal cycling. Thin-walled polypropylene construction delivers chemical compatibility and low sample loss, and plates are DNase-, RNase-, and pyrogen-free for sensitive molecular biology applications.
  • Well volume: 0.1 mL.
  • Wells: 96.
  • Skirt type: Semi skirt.
  • Color: White.
  • Notch position: A1 notch.
  • Pack quantity: 25 per pack.
  • Material: Polypropylene.
  • DNase- and RNase-free, and pyrogen free.
  • Autoclavable.
  • Typical application: PCR and thermal cycling workflows.