Learn More
Chemglass Life Sciences 0.1 mL PCR 8-Strip Tubes, Clear, Sterile (Nest)
SDP
Catalog No. 502733422
Encompass_Preferred
Low-profile 0.1 mL PCR 8-strip tubes in nest format, made from thin-walled polypropylene and supplied sterile. Designed for rapid heat transfer and low sample loss during thermal cycling, they are compatible with most thermal cyclers and real-time PCR instruments.
  • Thin-walled polypropylene for fast thermal transfer.
  • 0.1 mL capacity per tube.
  • 8-strip format in nest for high-throughput workflows.
  • Sterile, ready-to-use.
  • Leak-resistant easy-open caps minimize sample loss.
  • Compatible with most thermal cyclers and real-time PCR instruments.
Catalog No. 50-273-3422 Supplier Chemglass Life Sciences Supplier No. CGN3612C02
Low-profile 0.1 mL PCR 8-strip tubes in nest format, made from thin-walled polypropylene and supplied sterile. Designed for rapid heat transfer and low sample loss during thermal cycling, they are compatible with most thermal cyclers and real-time PCR instruments.
  • Thin-walled polypropylene for fast thermal transfer.
  • 0.1 mL capacity per tube.
  • 8-strip format in nest for high-throughput workflows.
  • Sterile, ready-to-use.
  • Leak-resistant easy-open caps minimize sample loss.
  • Compatible with most thermal cyclers and real-time PCR instruments.