Learn More
USA Scientific Inc 0.1 mL PCR 8-tube with attached flat caps, natural
Catalog No. NC0866066
Encompass
0.1 mL TempAssure PCR low profile 8-tube strips with individually attached, optically clear flat caps, natural. 120 strips of tubes with caps per pack.
Catalog No. NC0866066 Supplier USA Scientific Inc Supplier No. 14023700
0.1 mL TempAssure PCR low profile 8-tube strips with individually attached, optically clear flat caps, natural. 120 strips of tubes with caps per pack.