Learn More
USA Scientific Inc 0.1mL PCR 8-tube flat strip
Catalog No. 502745233
Encompass
0.1 mL TempAssure PCR flex-free natural color 8-tube strips with separate optically clear 8-cap (flat top) strips, natural. 125 strips each of tubes and caps per pack.
Catalog No. 50-274-5233 Supplier USA Scientific Inc Supplier No. 14023600
0.1 mL TempAssure PCR flex-free natural color 8-tube strips with separate optically clear 8-cap (flat top) strips, natural. 125 strips each of tubes and caps per pack.