Learn More
Thermo Scientific Dionex Ion Pairing Reagents | | 1 EA
Supplier:  Thermo Scientific 035360
Encompass
Type: Ion Pairing Reagent, Standard Concentration: 0.1 M, Operation: Used in mobile phase ion chromatography (MPIC) combining reversed-phase ion-pair chromatography with chemical suppression, Hazards: Corrosive, Standard Components: Tetrabutylammonium Hydroxide in Deionized Water, For Use With Application: MPIC, Volume (Metric): 500 mL, Storage: Room Temperature
Catalog No. NC0536288