Learn More
STEMCELL Technologies Heparin Solution, Size: 2 mL
SDP
Supplier:  STEMCELL Technologies 07980
Encompass
Heparin is a mucopolysaccharide with anticoagulant properties. It supports the binding of fibroblast growth factor (FGF) to its receptor and increases the stability of FGF. Heparin is used together with epidermal growth factor (EGF) and FGF in NeuroCult™ media for the culture of embryonic rat or adult human neural stem and progenitor cells, and adult mouse neural stem and progenitor cells. For complete instructions, refer to the Technical Manual: In Vitro Proliferation and Differentiation of Neural Stem and Progenitor Cells Using NeuroCult™ (Human; Document #28724 or Mouse/Rat; Document #28725), available at www.stemcell.com or contact us to request a copy. Heparin Solution is also required as a supplement in various other STEMCELL culture media, including MammoCult™ Human Medium Kit (Catalog #05620), EpiCult™-B Mouse Medium Kit (Catalog #05610), and EC-Cult™-XF Culture Kit (Catalog #08000).
Catalog No. NC0668440