Learn More
Bellco Glass, Inc. 0.2詢 Membrane S/Cap, 45mm
SDP
Catalog No. 501945760
Encompass_Preferred
45mm MEMBRANE S/CAPIt can be used for all bottles with gl45 thread & is characterized by: polypropylene materialA bore 3 holes in the center & welded in 0.2um ptfe foilA temperature resistance of up to 140°cAn extremely quick 2-way pressure compensation from 0.1 barUsage:Filling, packing, transporting & storing gas-generating mediaStoring critical substancesVarious biotechnological applications MADE IN THE USAFeatures:Extremely fast pressure compensation prevents both overpressure & vacuum. liquids, soil particles & solid matter cannot permeate into the bottleTo autoclave media - the membrane cap-unlie ordinary caps- is screwed tightly onto the bottle. There is no need for screwing & unscrewing the cap before or after the autoclaving process. The cap can be used over & again provided it is not damaged mechanically.
Catalog No. 50-194-5760 Supplier Bellco Glass, Inc. Supplier No. 563700046
45mm MEMBRANE S/CAPIt can be used for all bottles with gl45 thread & is characterized by: polypropylene materialA bore 3 holes in the center & welded in 0.2um ptfe foilA temperature resistance of up to 140°cAn extremely quick 2-way pressure compensation from 0.1 barUsage:Filling, packing, transporting & storing gas-generating mediaStoring critical substancesVarious biotechnological applications MADE IN THE USAFeatures:Extremely fast pressure compensation prevents both overpressure & vacuum. liquids, soil particles & solid matter cannot permeate into the bottleTo autoclave media - the membrane cap-unlie ordinary caps- is screwed tightly onto the bottle. There is no need for screwing & unscrewing the cap before or after the autoclaving process. The cap can be used over & again provided it is not damaged mechanically.