Learn More
Andwin Scientific 0.2 ML 8-TUBE STRIPS WITH INDI
SDP
Catalog No. 50819645
Encompass_Preferred
The 0.2 ml TempAssure PCR 8-tube strips feature individually attached dome caps that aid in eliminating the risk of cross-contamination. This product also has a thin wall that allow uniform and quick heat transfer for dependable applications. This 8-tube strip is guaranteed to be free of DNase, RNase, DNA, pyrogen, and PCR inhibitors.
This item is not returnable. View return policy
Catalog No. 50-819-645 Supplier Andwin Scientific Supplier No. 14022900
The 0.2 ml TempAssure PCR 8-tube strips feature individually attached dome caps that aid in eliminating the risk of cross-contamination. This product also has a thin wall that allow uniform and quick heat transfer for dependable applications. This 8-tube strip is guaranteed to be free of DNase, RNase, DNA, pyrogen, and PCR inhibitors.