Learn More
Andwin Scientific 0.2 ML 8-TUBE STRIPS WITH INDI
SDP
Supplier:  Andwin Scientific 14022908
Encompass_Preferred
The 0.2 mL 8-tube strips have dome caps that are individually attached, which prevents cross-contamination. Manufactured with thin walls, these tube strips can easily absorb heat for reliable amplifications. This product is free from DNase, RNase, DNA, and PCR. It comes in a pack of 120 strip in various colors of blue, green, red, violet, and yellow.
Catalog No. 50-819-646