Learn More
Andwin Scientific 0.2 ML 8-TUBE STRIPS WITH SEPA
SDP
Catalog No. 50819642
Encompass_Preferred
The 0.2 mL 8-tube strips are manufactured with thin walls, these tube strips can easily absorb heat for reliable amplifications. This product is free from DNase, RNase, DNA, and PCR. It comes in a pack of 125 strip in various colors of blue, green, violet, and yellow.
Catalog No. 50-819-642 Supplier Andwin Scientific Supplier No. 14022708
The 0.2 mL 8-tube strips are manufactured with thin walls, these tube strips can easily absorb heat for reliable amplifications. This product is free from DNase, RNase, DNA, and PCR. It comes in a pack of 125 strip in various colors of blue, green, violet, and yellow.