Learn More
Andwin Scientific 0.2 ML 96-PLACE STACKING PCR R
SDP
Catalog No. 50819913
Encompass_Preferred
The PCR tube rack can store skirted and non-skirted 96-well PCR plates or 0.2 mL tubes. This rack offers safe storage with the included clear lids that snap into base. This product is also stackable with or without the lid, allowing users to save space. It comes in a pack of various colors of blue, green, red, violet, and yellow.
Catalog No. 50-819-913 Supplier Andwin Scientific Supplier No. 23009708
The PCR tube rack can store skirted and non-skirted 96-well PCR plates or 0.2 mL tubes. This rack offers safe storage with the included clear lids that snap into base. This product is also stackable with or without the lid, allowing users to save space. It comes in a pack of various colors of blue, green, red, violet, and yellow.