Learn More
Diagenode Inc 0.2 ML MICROTUBES F/BIORUPTOR
Catalog No. NC1557052
Encompass
0.2 ml microtubes for Bioruptor® Pico1000/csThe 0.2 ml Bioruptor® Microtubes for DNA and chromatin shearing strongly improve DNA and chromatin sonication efficiency. These tubes have been thoroughly validated for use on the Bioruptor® Pico and are strongly recommended for DNA and chromatin shearing applications using this instrument. Recommended sample volume: 20-100 μl.
Catalog No. NC1557052 Supplier Diagenode Inc Supplier No. C30010020
0.2 ml microtubes for Bioruptor® Pico1000/csThe 0.2 ml Bioruptor® Microtubes for DNA and chromatin shearing strongly improve DNA and chromatin sonication efficiency. These tubes have been thoroughly validated for use on the Bioruptor® Pico and are strongly recommended for DNA and chromatin shearing applications using this instrument. Recommended sample volume: 20-100 μl.