Learn More
USA Scientific Inc 0.2 mL PCR 8-tube strip
Catalog No. 502745028
Encompass
0.2 mL TempAssure PCR 8-tube flex-free strips with individually attached, optically clear flat caps, violet. 120 strips of tubes with caps per pack.
Catalog No. 50-274-5028 Supplier USA Scientific Inc Supplier No. 14024705
0.2 mL TempAssure PCR 8-tube flex-free strips with individually attached, optically clear flat caps, violet. 120 strips of tubes with caps per pack.