Learn More
USA Scientific Inc 0.2 mL PCR 8-tube strip
Supplier:  USA Scientific Inc 14024701
Encompass
0.2 mL TempAssure PCR 8-tube flex-free strips with individually attached, optically clear flat caps, blue. 120 strips of tubes with caps per pack.
Catalog No. 50-274-5328