Learn More
Bio Rad Laboratories Trans-Blot Turbo RTA Midi 0.2 um Nitrocellulose Transfer Kit, for 40 blots
Supplier:  Bio Rad Laboratories 1704271
Encompass
Ready-to-assemble transfer kit includes 40 midi-sized 0.2 um nitrocellulose membranes (8.5 x 13.5 cm), 80 transfer stacks, 2 L 5x transfer buffer, and 2 gel trays for wetting and equilibrating membranes and transfer stacks
Catalog No. NC1797210
 Restricted Item