Learn More
Thermco Products Inc SPIRIT FILLED Thermometer, for ovens or oil baths
Catalog No. NC1389270
Encompass_Preferred
Thermco’s Special Application Thermometers are designed to be used with laboratory equipment. Calibrated against NIST standards to assure accuracy and reproducibility. Enlargements on the stem for proper immersions. Range: 1-200°C, Divisions: 1.0°C, Immersion: 125mm, Length: 405mm, No Safety Coat
Catalog No. NC1389270 Supplier Thermco Products Inc Supplier No. ACC639S
Thermco’s Special Application Thermometers are designed to be used with laboratory equipment. Calibrated against NIST standards to assure accuracy and reproducibility. Enlargements on the stem for proper immersions. Range: 1-200°C, Divisions: 1.0°C, Immersion: 125mm, Length: 405mm, No Safety Coat