Learn More
Chemglass Life Sciences Valve, Glass Portion Only, 1-Arm, 0-20mm Bore, 32mm OD Barrel, 22mm X 1.8W Sidearm
SDP
Catalog No. 501531784
Encompass_Preferred
CHEM-VAC™ valve glass portion only. Valve has a single sidearm sealed at a 90º angle to the barrel. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed. PLEASE NOTE: THE 0-20mm BORE SIZE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.
Catalog No. 50-153-1784 Supplier Chemglass Life Sciences Supplier No. CG50204
CHEM-VAC™ valve glass portion only. Valve has a single sidearm sealed at a 90º angle to the barrel. Heavy duty rounded coarse thread and precision formed tapered seat. Dimensions as listed. PLEASE NOTE: THE 0-20mm BORE SIZE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.