Learn More
Chemglass Life Sciences 0-20mm Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum
SDP
Catalog No. 501531792
Encompass_Preferred
0-20mm Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum. Replacement TFE plug and control knob for CHEM-VAC high vacuum valves CG-925 through CG-934. Plug is sold complete with control knob and TFE stem having three o-rings. PLEASE NOTE: CONTROL KNOB IS NOT SOLD SEPARATELY ON THE CG-900 SERIES VALVES. THE 0-20mm BORE SIZE VALVE IS Chemglass's PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.
Catalog No. 50-153-1792 Supplier Chemglass Life Sciences Supplier No. CG53504
0-20mm Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum. Replacement TFE plug and control knob for CHEM-VAC high vacuum valves CG-925 through CG-934. Plug is sold complete with control knob and TFE stem having three o-rings. PLEASE NOTE: CONTROL KNOB IS NOT SOLD SEPARATELY ON THE CG-900 SERIES VALVES. THE 0-20mm BORE SIZE VALVE IS Chemglass's PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.