Learn More
CELLTREAT Scientific Products SFCA Syringe Filter, 0.22m, 30mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229765
Encompass_Preferred
CELLTREAT Scientific Products 0.22¿m SFCA Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. SFCA (Surfactant Free Cellulose Acetate). Low protein binding, reduced extractables, and low hold-up volume. Filtration of aqueous solutions such as buffers, media, and reagents. Contains no pre-wetting agents. Membrane diameter 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister pack. 30/case.
Catalog No. 50-114-9064