Learn More
Chemglass Life Sciences Cap, GL-45, Gray, Vented, Non-Sterile, Polypropylene
SDP
Catalog No. 501215148
Encompass_Preferred
Polypropylene vented cap has a 0. 22um (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.They have a liner-less plug seal design for efficient sealing.They are autoclavable at 121 degreesC.
Catalog No. 50-121-5148 Supplier Chemglass Life Sciences Supplier No. CLS148410
Polypropylene vented cap has a 0. 22um (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination.They have a liner-less plug seal design for efficient sealing.They are autoclavable at 121 degreesC.