Learn More
Chemglass Life Sciences 0-25mm Drain Valve, One Piece, Spring Loaded
SDP
Catalog No. 5000980709
Encompass_Preferred
Replacement 0-25mm spring loaded drain valve complete with one-piece plug and control knob for temperature applications from -60 C to 230 C. Spring loaded drain valves compensate for temperature related material changes and are designed to provide a constant leak-free seal. For use with CG-1924-R, CG-1929-R, and CG-1930-R reaction vessels.
Catalog No. 50-009-80709 Supplier Chemglass Life Sciences Supplier No. CG911S251
Replacement 0-25mm spring loaded drain valve complete with one-piece plug and control knob for temperature applications from -60 C to 230 C. Spring loaded drain valves compensate for temperature related material changes and are designed to provide a constant leak-free seal. For use with CG-1924-R, CG-1929-R, and CG-1930-R reaction vessels.