Learn More
TRIBIOSCIENCE INC 0.2M Zinc Chloride Solution, 100 ML
SDP
Catalog No. NC3593062
Encompass
Zinc Chloride Solution is a formulated water solution. This ready-to-use solution can be used for protein structure and fold assays, nucleotide acid precipitation, and other biological analysis.
Catalog No. NC3593062 Supplier TRIBIOSCIENCE INC Supplier No. TBS5047
Zinc Chloride Solution is a formulated water solution. This ready-to-use solution can be used for protein structure and fold assays, nucleotide acid precipitation, and other biological analysis.