Learn More
Bachem Relaxin H2 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4039910.0200
Encompass_Preferred
Relaxin H2 (human) Trifluoroacetate, (Pyr-Leu-Tyr-Ser-Ala-Leu-Ala-Asn-Lys-Cys-Cys-His-Val-Gly-Cys-Thr-Lys-Arg-Ser-Leu-Ala-Arg-Phe-Cys-OH)A(H-Asp-Ser-Trp-Met-Glu-Glu-Val-Ile-Lys-Leu-Cys-Gly-Arg-Glu-Leu-Val-Arg-Ala-Gln-Ile-Ala-Ile-Cys-Gly-Met-Ser-Thr-Trp-Ser-OH)B, 99489-94-8, C256H408N74O74S8, Mr 5963.04, 0.2mg. Serelaxin. Relaxin H2 is the major stored and circulating isoform of the peptide hormone relaxin in humans. This pregnancy hormone has also shown cardioprotective activity. The observed long-term effects of relaxin H2 on connective tissues may be due to alterations in the turnover of collagen and proteoglycans.
Catalog No. NC1622152