Learn More
USA Scientific Inc 0.2 mL PCR 12-tube strip with 12-cap strips, mixed
Catalog No. NC9919403
Encompass
0.2 mL TempAssure PCR 12-tube strips with separate 12-cap (dome top) strips, mixed colors. 80 strips each of tubes and caps per pack.
Catalog No. NC9919403 Supplier USA Scientific Inc Supplier No. 14022408
0.2 mL TempAssure PCR 12-tube strips with separate 12-cap (dome top) strips, mixed colors. 80 strips each of tubes and caps per pack.