Learn More
USA Scientific Inc 0.2 mL PCR 8-tube strip with indiv. attached caps, mixed
Catalog No. NC9514330
Encompass
0.2 mL TempAssure PCR 8-tube strips with individually attached dome top caps, mixed. 120 strips of tubes with caps per pack. Polypropylene.
Catalog No. NC9514330 Supplier USA Scientific Inc Supplier No. 14022908
0.2 mL TempAssure PCR 8-tube strips with individually attached dome top caps, mixed. 120 strips of tubes with caps per pack. Polypropylene.