Learn More
Genesee Scientific Corporation Genesee Scientific 0.2 mL 8-Well PCR Strip Tubes, RNase/DNase Free, Optimized For Heat Transfer, 125 Strips/Unit
SDP
Catalog No. NC2400211
Encompass
Genesee Scientific 0.2 mL 8-Well PCR Strip Tubes are engineered for high-performance PCR applications. Made from ultra-clear, virgin polypropylene, they offer optimal heat transfer and rapid thermal cycling. Each tube is certified free from RNase, DNase, and other contaminants for reliable results. Featuring sturdy links for improved strip integrity and 45º angled cap hinges to minimize handling interference, these tubes are ideal for sensitive molecular biology workflows.
Catalog No. NC2400211 Supplier Genesee Scientific Corporation Supplier No. 27125
Genesee Scientific 0.2 mL 8-Well PCR Strip Tubes are engineered for high-performance PCR applications. Made from ultra-clear, virgin polypropylene, they offer optimal heat transfer and rapid thermal cycling. Each tube is certified free from RNase, DNase, and other contaminants for reliable results. Featuring sturdy links for improved strip integrity and 45º angled cap hinges to minimize handling interference, these tubes are ideal for sensitive molecular biology workflows.