Learn More
Andwin Scientific PCR 8-Well Tube Strips, 0.2 mL, 120/pk
SDP
Catalog No. NC1157020
Encompass_Preferred
Tube strips for real time PCR with individually attached flat caps. Ideal for all PCR protocols, these 0.1 and 0.2 mL tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
Catalog No. NC1157020 Supplier Andwin Scientific Supplier No. 93001118
Tube strips for real time PCR with individually attached flat caps. Ideal for all PCR protocols, these 0.1 and 0.2 mL tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.