Learn More
Alkali Scientific 0.2ml 96 Well PCR Plate, Semi Skirt, Clear, Black alpha numerics, 10/pk
SDP
Catalog No. 502389048
Encompass_Preferred
PCR PLATES* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%
Catalog No. 50-238-9048 Supplier Alkali Scientific Supplier No. PCNS96
PCR PLATES* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%