Learn More
Alkali Scientific 0.2mL 96 Well PCR Plate, Semi Skirt, White, 10/pk, 100/cs
SDP
Catalog No. 502388808
Encompass_Preferred
PCR PLATES* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%
Catalog No. 50-238-8808 Supplier Alkali Scientific Supplier No. PCN82
PCR PLATES* Made from polypropylene* Compatible with most thermal cyclers and real-time PCR instruments* The thin-walled design allows for high thermal conductivity to help reaction liquid reach the target temperature faster* Easy to open leak proof cap makes the loss of reaction less than 0.2%