Learn More
Opentrons Labworks Inc 0.2ML 96W PCR PLATE FULL 25PK
Catalog No. NC3346587
Encompass
Opentrons Tough 0.2 mL 96-Well PCR Plate, Full Skirt (25 count) Opentrons Tough 0.2 mL 96-Well PCR Plate, Full Skirt, RNase/DNase-free. Strong polycarbonate frame with thin-walled polypropylene wells for high-performance thermal cycling without warping. Ideal for use within automated liquid handlers with or without a gripper. 0.2 mL total well volume, 0.1 mL working volume. 25 count. Not for therapeutic use SP-2396
Catalog No. NC3346587 Supplier Opentrons Labworks Inc Supplier No. 99100076
Opentrons Tough 0.2 mL 96-Well PCR Plate, Full Skirt (25 count) Opentrons Tough 0.2 mL 96-Well PCR Plate, Full Skirt, RNase/DNase-free. Strong polycarbonate frame with thin-walled polypropylene wells for high-performance thermal cycling without warping. Ideal for use within automated liquid handlers with or without a gripper. 0.2 mL total well volume, 0.1 mL working volume. 25 count. Not for therapeutic use SP-2396