Learn More
Chemglass Life Sciences 0.2mL 96-Well PCR Plate, No Skirt, Clear, H1 Notch, 25/Pk
SDP
Catalog No. 501897742
Encompass_Preferred
PolypropylenePlate without skirt can be cut to any desired formatCut corner in position H1RNase/DNase/Pyrogen Free The low profile design minimizes the condensate contamination and evaporation of reaction solution
Catalog No. 50-189-7742 Supplier Chemglass Life Sciences Supplier No. CGN361501N
PolypropylenePlate without skirt can be cut to any desired formatCut corner in position H1RNase/DNase/Pyrogen Free The low profile design minimizes the condensate contamination and evaporation of reaction solution