Learn More
Chemglass Life Sciences 0.2mL 96-Well PCR Plate, White, No Skirt, H1 Notch, 25/Pack
SDP
Catalog No. 501897746
Encompass_Preferred
PolypropyleneWhite plates are ideal for qPCRNo side skirt so that it can be cut to any desired formatCut corner in position H1RNase/DNase/Pyrogen Free The low-profile design minimizes the condensate contamination and evaporation of reaction solution
Catalog No. 50-189-7746 Supplier Chemglass Life Sciences Supplier No. CGN3615W01N
PolypropyleneWhite plates are ideal for qPCRNo side skirt so that it can be cut to any desired formatCut corner in position H1RNase/DNase/Pyrogen Free The low-profile design minimizes the condensate contamination and evaporation of reaction solution