Learn More
Chemglass Life Sciences 0.2mL 96-Well PCR Plate, Semi-Skirt, Clear, A12 Notch - ABI Equivalent, 25/Pack
SDP
Catalog No. 501897744
Encompass_Preferred
PolypropylenePlate without skirt can be cut to any desired formatCut corner in position A12RNase/DNase/Pyrogen Free The low-profile design minimizes the condensate contamination and evaporation of reaction solution
Catalog No. 50-189-7744 Supplier Chemglass Life Sciences Supplier No. CGN361702
PolypropylenePlate without skirt can be cut to any desired formatCut corner in position A12RNase/DNase/Pyrogen Free The low-profile design minimizes the condensate contamination and evaporation of reaction solution