Learn More
USA Scientific Inc 0.2 mL TempAssure PCR tube, attached flat cap, mixed
Catalog No. NC9642455
Encompass_Preferred
0.2 mL TempAssure PCR tube, attached flat cap, mixed colors. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag. Polypropylene.
Catalog No. NC9642455 Supplier USA Scientific Inc Supplier No. 14028108
0.2 mL TempAssure PCR tube, attached flat cap, mixed colors. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag. Polypropylene.