Learn More
Andwin Scientific 0.2ML FLAT CAP PCR TUBE 1000PK
SDP
Catalog No. NC0096149
Encompass_Preferred
The 0.2 mL PCR Tubes and Caps is ideal for use in all PCR procedures. This product can be utilized with most popular thermal cycler blocks. The tubes are autoclavable and are chemically inert. Its smooth, thin, uniform walls permit correct thermal tr
Catalog No. NC0096149 Supplier Andwin Scientific Supplier No. 20170012
The 0.2 mL PCR Tubes and Caps is ideal for use in all PCR procedures. This product can be utilized with most popular thermal cycler blocks. The tubes are autoclavable and are chemically inert. Its smooth, thin, uniform walls permit correct thermal tr