Learn More
Chemglass Life Sciences PCR Reaction 8-Tube Strips, 0.2 mL, Clear, Attached Flat Caps
SDP
Catalog No. 502733165
Encompass_Preferred
Thin-wall, clear 0.2 mL PCR 8-tube strips with attached flat caps that provide secure sealing and efficient thermal transfer for PCR cycling.
  • 0.2 mL capacity per tube.
  • Eight tubes per strip.
  • Attached flat caps for secure sealing and easy opening.
  • Extra-thin wall for optimal thermal transfer and reduced cycle times.
  • Clear polypropylene construction for optical clarity.
  • Supplied in resealable bags; non-sterile and autoclavable.
Catalog No. 50-273-3165 Supplier Chemglass Life Sciences Supplier No. CLS3611V01
Thin-wall, clear 0.2 mL PCR 8-tube strips with attached flat caps that provide secure sealing and efficient thermal transfer for PCR cycling.
  • 0.2 mL capacity per tube.
  • Eight tubes per strip.
  • Attached flat caps for secure sealing and easy opening.
  • Extra-thin wall for optimal thermal transfer and reduced cycle times.
  • Clear polypropylene construction for optical clarity.
  • Supplied in resealable bags; non-sterile and autoclavable.